missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ National Target™ Empty Caps without Septum for 10-425 Vials
Click to view available options
Color:
Green
Red
Yellow
Unit Size:
Pack of 100
Description
Thermo Scientific™ National Target Empty Open-Top Caps for 10-425 Vials are designed to be used with any of our 10mm septa.
- Polypropylene caps are chemically inert and suitable for most chromatography applications
- Target 10-425 caps feature a flanged rim, making them suitable for Japanese autosampler trays
Specifications
Specifications
| Description | Green; For 10-425 Target Vials |
| Color | Green |
| Material | Polypropylene |
| Product Line | National |
| Closure Size | 10-425 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction