Learn More
Thermo Scientific™ National Target DP™ Screw Caps with Septa
Click to view available options
Color:
Black
Blue
Green
Ivory, Red
Ivory, Red, Blue
Ivory, Red, Yellow
Red
Red, Black
Red, Green
Yellow
Septum:
Blue PTFE/Silicone
Ivory PTFE/Red Rubber
PTFE/Silicone
PTFE/Silicone/PTFE
Unit Size:
Pack of 100
Description
Thermo Scientific™ National Target DP Screw Caps with Septa are color coded to separate assays, standards or samples.
- Preassembled with septa
Specifications
Specifications
| Description | Ivory PTFE/Red Rubber Septa; Blue |
| Color | Ivory, Red, Blue |
| Material | PTFE, Rubber |
| Septum | Ivory PTFE/Red Rubber |
| Product Line | National Target DP |
| Closure Size | 9-425 |