missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Snap-It™ Closures for 12 x 32 crimp/snap vials

Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.

Brand:  Thermo Scientific™ C4011-53G

Code : 50

Additional Details : Weight : 0.29000kg

 View more versions of this product

Product Code. 13150963

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.

  • Caps eliminate the need for crimping or de-capping tools
  • Easy to apply and easy to remove from vials
  • Available in a variety of colors and septa choices
Specifications

Specifications

C4011-53G
PTFE, Silicone
Red PTFE/White Silicone/Red PTFE
11 mm
Green
2 mL
National Snap-It
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers