Learn More
Thermo Scientific™ National Preassembled Target DP™ Kits
Product Code. p-4526330
Click to view available options
Closure Color:
Blue
Red
Septum:
PTFE/Rubber
PTFE/Silicone
PTFE/Silicone/PTFE
Color:
Amber
Clear
Unit Size:
Pack of 100
Product Code. 11901593
Brand: Thermo Scientific™ C4000180W
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
  • Packaged in convenient vial trays with clear covers or in polybags
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Color (Seal) Red
Description PTFE/red rubber septa; Clear glass I-D
Color Clear
Material Glass
Volume (Metric) 2 mL
Septum PTFE/Rubber
Closure Size 9-425
Marking Spot Yes