Learn More
Thermo Scientific™ National Polypropylene and Glastic™ Vials with 8-425 Finish
Product Code. p-4526449
Change view
Click to view available options
Volume (Metric) Working:
300 μL
400 μL
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Working unitSize
12440108 300 μL Pack of 100
12490108 400 μL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12440108 Supplier Thermo Scientific™ Supplier No. C401315GST
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
  • Precision formed tips are designed to aid sample recovery for limited-volume applications
  • Glastic vial features a glass insert pre-inserted inside a clear TPX vial
TRUSTED_SUSTAINABILITY
Specifications
Description 150µL Glastic Glass Insert/ TPX MicroVials
Color Clear
Material Glass, TPX
Volume (Metric) 450 μL
Volume (Metric) Residual <2 μL
Volume (Metric) Working 300 μL
Quantity 100/Pk.
Closure Size 8-425