Learn More
Thermo Scientific™ National Inserts for Standard-Opening Vials
Product Code. 12430098
Change view
Click to view available options
Volume (Metric):
200 μL
225 μL
250 μL
Unit Size:
Pack of 100
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12430098 200 μL Pack of 100
10688995 225 μL Pack of 100
12480158 250 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12430098 Supplier Thermo Scientific™ Supplier No. C4012K530
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
  • Pulled point inserts are an economical choice for non-critical applications
TRUSTED_SUSTAINABILITY
Specifications
Description C4012-K530
Material Kimshield Glass
Volume (Metric) 200 μL
Style Polyspring, Pulled point
Base Style Conical
Product Line National
Product Type Insert