missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Economy Vials

Product Code. 11573830 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Amber
Clear
Product Type:
Clear
Crimp Top Vial
Screw Thread Vial
Snap cap
Volume (Metric) Working:
2 mL
Unit Size:
Pack of 1000
Product Code. Color Product Type Volume (Metric) Working unitSize
11573830 Clear Crimp Top Vial 2 mL Pack of 1000
11583830 Amber Crimp Top Vial 2 mL Pack of 1000
11593830 Clear Clear 2 mL Pack of 1000
13287489 Amber Snap cap 2 mL Pack of 1000
10003873 Clear Screw Thread Vial 2 mL Pack of 1000
11702197 Amber Screw Thread Vial 2 mL Pack of 1000
11563860 Clear Screw Thread Vial 2 mL Pack of 1000
11573860 Amber Screw Thread Vial 2 mL Pack of 1000
8 options
This item is not returnable. View return policy

Product Code. 11573830

Brand: Thermo Scientific™ 60180701

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ National Economy Vials are offered as clear or amber, in a range of styles to meet specific application needs.

Thermo Scientific™ National Economy Vials are offered as clear or amber, in a range of styles to meet specific application needs.

TRUSTED_SUSTAINABILITY

Specifications

Description 60180-701
Color Clear
Volume (Metric) 2 mL
Volume (Metric) Working 2 mL
For Use With (Equipment) Thermo Scientific, Agilent, PE, Shimadzu, Varian
Product Type Crimp Top Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.