missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Components for Vial Convenience Kits

Product Code. 12478646
Change view
Click to view available options
Unit Size:
Pack of 100
Volume (Metric):
20 mL
Color:
Amber
Clear
Product Code. unitSize Volume (Metric) Color
12478646 Pack of 100 - -
12408646 Pack of 100 20 mL Clear
12438646 Pack of 100 20 mL Amber
3 options
This item is not returnable. View return policy

Product Code. 12478646

Brand: Thermo Scientific™ B795026

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Vials, caps and septa for use with Thermo Scientific™ National Vial Convenience Kits.

Vials, caps and septa for use with Thermo Scientific™ National vial convenience kits.

TRUSTED_SUSTAINABILITY

Specifications

Description Septa Only
Material PTFE, Silicone (Septa)
Closure Material PTFE, Silicone
Quantity 100 Pk.
For Use With (Equipment) Thermo Scientific™ National Vial Convenience Kits
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Product Type Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.