Learn More
Thermo Scientific™ National Assembled Convenience Kits: EPA Vials with Caps and Septa
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
£136.67 - £145.00
Product Code | Brand | Volume (Metric) | Color | Certifications/Compliance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Certifications/Compliance | Price | Quantity & Availability | |||
12498646
|
Thermo Scientific™
B7950-C |
40 mL | Clear | Class 200 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12408656
|
Thermo Scientific™
B7950-C3 |
40 mL | Clear | Class 300 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
- Recommended for discrete water sampling under EPA 40 CFR 136 “Guidelines for Establishing Test Procedures for the Analysis of Pollutants” and EPA 40 CFR 141 “National Interim Primary Drinking Water Regulations: Control of Trihalomethanes in Drinking Water”
- Class 100—Vials, Caps and Septa are assembled
- Class 200—Vials, Caps and Septa are processed to EPA specifications and assembled
- Class 300—Vials, Caps and Septa are processed and certified to EPA specifications, then assembled (Certificate of Analysis included with each box of vials)
Specifications
Discrete Water Sampling Applications |