missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nateglinide
GREENER_CHOICE

Product Code. 18782714
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18782714

Brand: Tocris Bioscience™ 4231/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kir6 (KATP) blocker; displays high affinity for SUR1/Kir6.2 channels

Kir6 (KATP) blocker. Exhibits in vitro tissue selectivity for pancreatic β-cell-type Kir6 channels over cardiovascular Kir6 channels; displays high affinity for SUR1/Kir6.2 channels. Hypoglycemic agent; stimulates insulin secretion from pancreatic β-cells by increasing cytosolic Ca2+ concentration. Also agonist at the orphan receptor MRGPRX4.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105816-04-4
Molecular Formula C19H27NO3

Specifications

Quantity 10 mg
Formula Weight 317.42
Purity >98%
Molecular Weight (g/mol) 317.429
Chemical Name or Material Nateglinide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.