missing translation for 'onlineSavingsMsg'
Learn More

Nateglinide Methyl Ester, Mikromol™

Product Code. 30049812
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049812 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049812 Supplier Mikromol Supplier No. MM0847.100025
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Nateglinide Methyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nateglinide Methyl Ester
CAS 105746-47-2
Analyte or Component Names Nateglinide Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C20 H29 N O3
Certifications/Compliance ISO 9001
Synonym Nateglinide Methyl Ester, N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalanine methyl ester, D-Phenylalanine, N-[[4-(1-methylethyl)cyclohexyl]carbonyl]-, methyl ester, trans-
InChI Formula InChI=1S/C20H29NO3/c1-14(2)16-9-11-17(12-10-16)19(22)21-18(20(23)24-3)13-15-7-5-4-6-8-15/h4-8,14,16-18H,9-13H2,1-3H3,(H,21,22)/t16-,17-,18-/m1/s1
SMILES COC(=O)[C@@H](Cc1ccccc1)NC(=O)[C@@H]2CC[C@H](CC2)C(C)C
IUPAC Name methyl (2R)-3-phenyl-2-[(4-propan-2-ylcyclohexanecarbonyl)amino]propanoate
Molecular Weight (g/mol) 331.45
Quantity 25 mg
Formula Weight 331.2147 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.