missing translation for 'onlineSavingsMsg'
Learn More

Nateglinide Methyl Ester Impurity, TRC

Product Code. 30482856
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482856 Supplier Toronto Research Chemicals Supplier No. TRCN379410100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nateglinide Methyl Ester
CAS 105746-47-2
Molecular Formula C20 H29 N O3
Quantity 100 mg
Synonym N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalanine methyl ester, D-Phenylalanine','N-[[4-(1-methylethyl)cyclohexyl]carbonyl]-','methyl ester','trans-
InChI Formula InChI=1S/C20H29NO3/c1-14(2)16-9-11-17(12-10-16)19(22)21-18(20(23)24-3)13-15-7-5-4-6-8-15/h4-8,14,16-18H,9-13H2,1-3H3,(H,21,22)/t16-,17-,18-/m1/s1
SMILES COC(=O)[C@@H](Cc1ccccc1)NC(=O)[C@@H]2CC[C@H](CC2)C(C)C
IUPAC Name methyl (2R)-3-phenyl-2-[(4-propan-2-ylcyclohexanecarbonyl)amino]propanoate
Molecular Weight (g/mol) 331.45
Formula Weight 331.2147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.