missing translation for 'onlineSavingsMsg'
Learn More

Nateglinide, MedChemExpress

Product Code. 16408488
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16408488

Brand: Medchem Express HYB0422

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nateglinide, a D-phenylalanine derivative, is an orally active and short-acting insulinotropic agent and a DPP IV inhibitor. Nateglinide inhibits ATP-sensitive K+ channels in pancreatic β-cells. Nateglinide is used for the treatment of type 2 (non-insulin-dependent) diabetes mellitus.

Nateglinide, a D-phenylalanine derivative, is an orally active and short-acting insulinotropic agent and a DPP IV inhibitor. Nateglinide inhibits ATP-sensitive K+ channels in pancreatic β-cells. Nateglinide is used for the treatment of type 2 (non-insulin-dependent) diabetes mellitus.

Specifications

Chemical Name or Material Nateglinide
Physical Form Solid
CAS 105816-04-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H27NO3
For Use With (Application) Neuroscience-Neuromodulation
Synonym A4166 Senaglinide
Solubility Information DMSO : 100 mg/mL (315.04 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](CC1=CC=CC=C1)NC([C@H]2CC[C@H](C(C)C)CC2)=O
Molecular Weight (g/mol) 317.42
Formula Weight 317.42
Percent Purity 99.78%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.