missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAT9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576484
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
NAT-9 (N-acetyltransferase 9), also known as EBSP (embryo brain-specific protein), is a 207 amino acid protein belonging to the acetyltransferase family and the GNAT subfamily. Containing a N-acetyltransferase domain, NAT-9 may be associated with psoriasis and psoriatic arthritis, a type of inflammatory/autoimmune disease that affects skin, tendons and/or joints of the hands and feet. Expressed as two isoforms produced by alternative splicing events, NAT-9 is encoded by a gene located on human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes.
Specifications
| NAT9 | |
| Polyclonal | |
| Unconjugated | |
| NAT9 | |
| EBS; EBSP, hNATL; embryo brain specific protein; Embryo brain-specific protein; N-acetyltransferase 9; N-acetyltransferase 9 (GCN5-related, putative); N-acetyltransferase 9 (putative); NAT9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26151 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BTE0 | |
| NAT9 | |
| Recombinant fusion protein of human NAT9. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction