missing translation for 'onlineSavingsMsg'
Learn More

NAT8L Antibody, Novus Biologicals™

Product Code. 18489940 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18489940 0.1 mL 0.1mL
18407690 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18489940 Supplier Novus Biologicals Supplier No. NBP183479

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NAT8L Polyclonal antibody specifically detects NAT8L in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT8L
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias Camello-like protein 3, CML3, DKFZp434B0650, DKFZp566H184, EC 2.3.1.17, FLJ37478, Hcml3, MGC117272, NAA synthetase, N-acetyltransferase 8-like, N-acetyltransferase 8-like (GCN5-related, putative), N-acetyltransferase 8-like protein, NAT8-LIKE
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:MADIEQYYMKPPGSCFWVAVLDGNVVGIVAARAHEEDNTVELLRMSVDSRFRGKGIAKALGRKVLE
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339983
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.