missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAT2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145439
This item is not returnable.
View return policy
Description
Participates in the detoxification of a plethora of hydrazine and arylamine drugs. Catalyzes the N- or O-acetylation of various arylamine and heterocyclic amine substrates and is able to bioactivate several known carcinogens.
Specifications
| NAT2 | |
| Polyclonal | |
| Unconjugated | |
| NAT2 | |
| Aac2; Arylamide acetylase 2; arylamine N-acetyltransferase 2; AT-2; AV377607; N-acetyl transferase 2; N-acetyltransferase 2; N-acetyltransferase 2 (arylamine N-acetyltransferase); N-acetyltransferase type 2; N-Acetyltransferase-2 (arylamine N-acetyltransferase); Nat2; NAT-2; Nat2a; PNAT; Polymorphic arylamine N-acetyltransferase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10, 116632, 17961 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P11245, P50295, P50298 | |
| NAT2 | |
| Recombinant Rat NAT2 protein, Asp20-Val280 (Accession #P50298). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction