missing translation for 'onlineSavingsMsg'
Learn More

NAT13 Antibody, Novus Biologicals™

Product Code. 18349485 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349485 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349485 Supplier Novus Biologicals Supplier No. NBP318423

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NAT13 Polyclonal antibody specifically detects NAT13 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT13
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 20 ug/mg lysate
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias EC 2.3.1.-, FLJ13194, hSAN, Mak3 homolog (S. cerevisiae), N(alpha)-acetyltransferase 50, NatE catalytic subunit, N-acetyltransferase 13 (GCN5-related), N-acetyltransferase 5, N-acetyltransferase san homolog, NAT13San, NAT5SAN, NatE catalytic subunit, separation anxiety
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 119 and 169..
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 80218
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.