missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAT11 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-77095CL1
This item is not returnable.
View return policy
Description
NAT11 Polyclonal antibody specifically detects NAT11 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| NAT11 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| EC 2.3.1.-, FLJ13848, N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae), N-acetyltransferase 11, N-acetyltransferase 11 (GCN5-related, putative), N-alpha-acetyltransferase 40, NatD catalytic subunit, NAT11, PATT1 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human NAT11. The immunogen is located within the last 50 amino acids of NAT11. Amino Acid Squence: YEILSRRTKFGDSHH | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology | |
| 79829 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction