missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAT10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577193
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
IRS4 encodes the insulin receptor substrate 4, a cytoplasmic protein that contains many potential tyrosine and serine/threonine phosphorylation sites. Tyrosine-phosphorylated IRS4 protein has been shown to associate with cytoplasmic signalling molecules that contain SH2 domains. The IRS4 protein is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation.
Specifications
| NAT10 | |
| Polyclonal | |
| Unconjugated | |
| Nat10 | |
| 18S rRNA cytosine acetyltransferase; AI429152; ALP; hALP; KIAA1709; N-acetyltransferase 10; N-acetyltransferase 10 (GCN5-related); N-acetyltransferase-like protein; NAT10; NET43; novel putative ATPase domain containing protein; RGD1306717; RNA cytidine acetyltransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 311257, 55226 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H0A0 | |
| Nat10 | |
| Recombinant full length Human NAT10. | |
| 100 μL | |
| Primary | |
| Human, Rat, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction