missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAT10 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32263V
This item is not returnable.
View return policy
Description
NAT10 Polyclonal antibody specifically detects NAT10 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| NAT10 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| ALP, DKFZp434C116, EC 2.3.1.-, FLJ10774, FLJ12179, FLJ23850, hALP, KIAA1709, N-acetyltransferase 10, N-acetyltransferase 10 (GCN5-related), N-acetyltransferase-like protein, NET43 | |
| The immunogen this antibody was made to, maps to a region between residue 650 to 700 of human N-acetyltransferase 10 using the numbering given in entry NP_078938.2 (GeneID 55226). | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Direct Reversal of DNA Damage, Immunology, Signal Transduction | |
| 55226 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction