Learn More
Invitrogen™ NAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595422
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: A431 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Arylamine N-acetyltransferases (NAT-1 and NAT-2) catalyze N- or O-acetylation of heterocyclic and arylamine substrates in the detoxification of a wide array of drugs. Certain alleles causing high N-acetyltransferase activity have been associated with colon and urinary bladder cancers, as NATs also bioactivate several known carcinogens. Both NAT-1 and NAT-2 are cytoplasmic proteins and play an active role in the detoxification of many arylamine and hydrazine drugs. N-acetylation polymorphism is determined by the level of NAT activity in liver tissues, and has been linked to the action and toxicity of drugs that contain amines. Human NAT-1 is the functional homolog of rodent NAT-2, while human NAT-2 is the functional homolog of rodent NAT-1.
Specifications
| NAT1 | |
| Polyclonal | |
| Unconjugated | |
| NAT1 | |
| AAC1; Arylamide acetylase 1; arylamine N-acetyltransferase 1; AT-1; MNAT; Monomorphic arylamine N-acetyltransferase; N(alpha)-acetyltransferase 15, NatA auxiliary subunit; N-acetyl transferase 1; N-acetyl transferase 1, liver, blood; N-acetyltransferase (arylamine N-acetyltransferase); N-acetyltransferase 1; N-acetyltransferase 1 (arylamine N-acetyltransferase); N-acetyltransferase type 1; Nat; Nat1; NAT-1; NATI | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P18440 | |
| NAT1 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human NAT1 (172-201aa NEEFLHSDLLEDSKYRKIYSFTLKPRTIED). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.