missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NAT1 Antibody (SR4177), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27820-100ul
This item is not returnable.
View return policy
Description
NAT1 Monoclonal antibody specifically detects NAT1 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| NAT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR4177 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| AAC1N-acetyltransferase type 1, Arylamide acetylase 1, arylamine N-acetyltransferase 1, EC 2.3.1.5, MNAT, Monomorphic arylamine N-acetyltransferase, N-acetyltransferase 1 (arylamine N-acetyltransferase), NAT-1, NATI | |
| A synthesized peptide derived from human NAT1 (Uniprot #: P18440) | |
| 100 μL | |
| metabolism | |
| 9 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction