missing translation for 'onlineSavingsMsg'
Learn More

Naspm, MedChemExpress

Product Code. 16497937
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497937

Brand: Medchem Express HY12506

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Naspm (1-Naphthyl acetyl spermine), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.

Naspm (1-Naphthyl acetyl spermine), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.

Specifications

Chemical Name or Material Naspm
Physical Form Solid
CAS 122306-11-0
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H34N4O
For Use With (Application) Neuroscience-Neuromodulation
Synonym 1-Naphthylacetyl spermine
Solubility Information DMSO : 10 mg/mL (26.99 mM; Need ultrasonic)
SMILES O=C(NCCCNCCCCNCCCN)CC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 370.53
Formula Weight 370.53
Percent Purity 95.18%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.