missing translation for 'onlineSavingsMsg'
Learn More

NASP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-38352-100ul

 View more versions of this product

Product Code. 30226783

  • £417.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NASP Polyclonal antibody specifically detects NASP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NASP
Polyclonal
Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
DKFZp547F162, FLB7527, FLJ31599, FLJ35510, histone H1-binding protein, MGC19722, MGC20372, MGC2297, nuclear autoantigenic sperm protein, nuclear autoantigenic sperm protein (histone-binding), PRO1999
Recombinant fusion protein containing a sequence corresponding to amino acids 180-449 of human NASP (NP_689511.2).,, Sequence:, WDMLDLAKIIFKRQETKEAQLYAAQAHLKLGEVSVESENYVQAVEEFQSCLNLQEQYLEAHDRLLAETHYQLGLAYGYNSQYDEAVAQFSKSIEVIENRMAVLNEQVKEAEGSSAEYKKEIEELKELLPEIREKIEDAKESQRSGNVAELALKATLVESSTSGFTPGGGGSSVSMIASRKPTDGASSSNCVTDISHLVRKKRKPEEESPRKDDAKKAKQEPEVNGGSGDAVPSGNEVSENMEEEAENQAESRAAVEGTVEAGATVESTAC
100 μL
Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, modENCODE Antibodies, Stem Cell Markers
4678
Store at -20°C. Avoid freeze-thaw cycles.
IgG
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.