missing translation for 'onlineSavingsMsg'
Learn More

NASP Antibody, DyLight 594, Novus Biologicals™

Product Code. 18399711 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399711 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399711 Supplier Novus Biologicals Supplier No. NBP306445DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NASP Polyclonal antibody specifically detects NASP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NASP
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias DKFZp547F162, FLB7527, FLJ31599, FLJ35510, histone H1-binding protein, MGC19722, MGC20372, MGC2297, nuclear autoantigenic sperm protein, nuclear autoantigenic sperm protein (histone-binding), PRO1999
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NASP fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, modENCODE Antibodies, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4678
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.