missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAS1/SLC13A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25039-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| NAS1/SLC13A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC13A1 | |
| hNaSi 1, Na(+)/sulfate cotransporter, NAS1, NaSi 1, NASI1, SLC13A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55961, 6561 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BZW2, Q9JHI4 | |
| Slc13a1 | |
| NAS1/SLC13A1 Fusion Protein Ag19026 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction