missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NARS2 Antibody (OTI10C10), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72876UV
This item is not returnable.
View return policy
Description
NARS2 Monoclonal specifically detects NARS2 in Human samples. It is validated for Western Blot.
Specifications
| NARS2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NARS2 | |
| Full length recombinant protein of human NARS2 (NP_078954) produced in HEK293T cells. | |
| 0.1 mL | |
| 79731.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI10C10 | |
| Western Blot | |
| asnRS, asparagine tRNA ligase 2, mitochondrial (putative), Asparagine--tRNA ligase, asparaginyl-tRNA synthetase 2, mitochondrial (putative), EC 6.1.1, EC 6.1.1.22, FLJ23441, probable asparaginyl-tRNA synthetase, mitochondrial, SLM5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction