Learn More
Invitrogen™ NARS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116533
Description
Positive Samples: HT-29, 293T, LO2, Jurkat, Mouse pancreas Immunogen sequence: PALGDVFCIA QSYRAEQSRT RRHLAEYTHV EAECPFLTFD DLLNRLEDLV CDVVDRILKS PAGSIVHELN PNFQPPKRPF KRMNYSDAIV WLKEHDVKKE DGTFYEFGED IPEAPERLMT DTINEPILLC RFPVEIKSFY MQRCPEDSRL TESVDVLMPN VGEIVGGSMR IFDSEEILAG YKREGIDPTP YYWYTDQRKY GTCPHGGYGL GLERFLTWIL NRYHIRDVCL YPRFVQRCTP.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Asparaginyl-tRNA synthetase is localized to the cytoplasm and belongs to the class II family of tRNA synthetases. The N-terminal domain represents the signature sequence for the eukaryotic asparaginyl-tRNA synthetases.
Specifications
| NARS | |
| Polyclonal | |
| Unconjugated | |
| NARS1 | |
| 3010001M15Rik; AA960128; AsnRS; asparagine tRNA ligase 1, cytoplasmic; asparagine--tRNA ligase, cytoplasmic; Asparaginyl-tRNA synthetase; asparaginyl-tRNA synthetase, cytoplasmic; C78150; LRRGT00113; Nars; NARS1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4677, 70223 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.98 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O43776, Q8BP47 | |
| NARS1 | |
| Recombinant protein (or fragment)). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.