missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Soda Lime Glass Reagent Bottles
Click to view available options
Capacity (Metric):
30 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Color:
Amber
Clear
Unit Size:
Pack of 10
Pack of 2
Description
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Amber |
| Includes | Interchangeable octagonal head, Polypropylene stopper |
| Material | Borosilicate Glass |
| No. per Pack | 10 |
| Product Type | Reagent Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction