missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hecht Karl™ Assistant™ Amber Reagent Bottle With Glass Stopper
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Each
Description
- Amber glass
- Conical shoulder
- Ground glass stopper
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Color | Amber |
| Description | Amber glass reagent bottle, conical shoulder, ground glass stopper |
| Form | Conical shoulder |
| Material | Glass |
| Sterility | Non-sterile |
| Stopper Number | 14.5 |
| Closure Color | Amber |
| Closure Included | Yes |
| Graduated | No |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction