missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naringin Dihydrochalcone, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Naringin Dihydrochalcone |
| CAS | 18916-17-1 |
| Molecular Formula | C27H34O14 |
| Quantity | 25 g |
| Synonym | beta-D-3,5-Dihydroxy-4-(p-hydroxyhydrocinnamoyl)phenyl 2-O-(6-deoxy-alpha-L-mannopyranosyl)glucopyranoside, Phloretin','4'-(2-O-alpha-L-rhamno-beta-D-glucopyranoside), Naringin Dihydrochalcone |
| InChI Formula | InChI=1S/C27H34O14/c1-11-20(33)22(35)24(37)26(38-11)41-25-23(36)21(34)18(10-28)40-27(25)39-14-8-16(31)19(17(32)9-14)15(30)7-4-12-2-5-13(29)6-3-12/h2-3,5-6,8-9,11,18,20-29,31-37H,4,7,10H2,1H3/t11-,18+,20-,21+,22+,23-,24+,25+,26-,27+/m0/s1 |
| SMILES | C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c(C(=O)CCc4ccc(O)cc4)c(O)c3)[C@H](O)[C@H](O)[C@H]1O |
| IUPAC Name | 1-[4-[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-2,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)propan-1-one |
| Molecular Weight (g/mol) | 582.55 |
| Formula Weight | 582.1949 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction