missing translation for 'onlineSavingsMsg'
Learn More

Naringin Dihydrochalcone, MedChemExpress

Product Code. 16482346
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 16482346

Brand: Medchem Express HYN0119

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Naringin Dihydrochalcone is an artificial sweetener derived from naringin. Naringin is a major flavanone glycoside obtained from tomatoes, grapefruits, and many other citrus fruits. Naringin exhibits biological properties such as antioxidant, anti-inflammatory, and antiapoptotic activities. Naringin suppresses NF-κB signaling pathway.

Naringin Dihydrochalcone is an artificial sweetener derived from naringin. Naringin is a major flavanone glycoside obtained from tomatoes, grapefruits, and many other citrus fruits. Naringin exhibits biological properties such as antioxidant, anti-inflammatory, and antiapoptotic activities. Naringin suppresses NF-κB signaling pathway.

Specifications

Chemical Name or Material Naringin Dihydrochalcone
CAS 18916-17-1
Color White
Purity Grade Notes Research
Molecular Formula C27H34O14
Quantity 500 mg
Synonym Naringin DC
Solubility Information DMSO : ≥ 100 mg/mL (171.66 mM)
SMILES O=C(C1=C(O)C=C(O[C@H]2[C@@H]([C@H]([C@@H]([C@@H](CO)O2)O)O)O[C@@]3([H])[C@@H]([C@@H]([C@H]([C@H](C)O3)O)O)O)C=C1O)CCC4=CC=C(O)C=C4
Molecular Weight (g/mol) 582.55
Formula Weight 582.55
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.