missing translation for 'onlineSavingsMsg'
Learn More

Naringin dihydrochalcone, 98%, Thermo Scientific Chemicals

Product Code. 15448349
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15448349

Brand: Thermo Scientific Alfa Aesar J66830.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18916-17-1
Molecular Formula C27H34O14
Molecular Weight (g/mol) 582.555
MDL Number MFCD08436145
InChI Key CWBZAESOUBENAP-NDAHTVSZSA-N
Synonym 1-[4-[(2-O-(6-Deoxy-a-L-rhamnopyranosyl)-b-D-glucopyranosyl]oxy]-2,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)-1-propanone
PubChem CID 126963586
IUPAC Name 1-[4-[(3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-2,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)propan-1-one
SMILES CC1C(C(C(C(O1)OC2C(C(C(OC2OC3=CC(=C(C(=C3)O)C(=O)CCC4=CC=C(C=C4)O)O)CO)O)O)O)O)O

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in DMSO
Formula Weight 582.55
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Naringin dihydrochalcone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.