missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NARFL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13652-1-AP-150UL
This item is not returnable.
View return policy
Description
Component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into extramitochondrial Fe/S proteins. Seems to negatively regulate the level of HIF1A expression, although this effect could be indirect.Specifications
| NARFL | |
| Polyclonal | |
| Unconjugated | |
| NARFL | |
| HPRN, IOP1, LET1L, NARFL, PRN, Protein related to Narf | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 360496, 64428, 67563 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5BK18, Q7TMW6, Q9H6Q4 | |
| Narfl | |
| NARFL Fusion Protein Ag4217 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction