missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NARFL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106695
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NARFL.
Component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into extramitochondrial Fe/S proteins. Seems to negatively regulate the level of HIF1A expression, although this effect could be indirect.
Specifications
| NARFL | |
| Polyclonal | |
| Unconjugated | |
| Narfl | |
| 9030612I22Rik; AI504405; CIAO3; Cytosolic Fe-S cluster assembly factor NARFL; Cytosolic iron-sulfur assembly component 3; HPRN; IOP1; iron-only hydrogenase-like protein 1; LET1 like/JFP15; LET1L; NAR1; NARFL; nuclear prelamin A recognition factor like; nuclear prelamin A recognition factor-like; nuclear prelamin A recognition factor-like protein; PRN; protein related to Narf | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 64428 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H6Q4 | |
| Narfl | |
| A synthesized peptide derived from human NARFL(Accession Q9H6Q4), corresponding to amino acid residues Q231-L281. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction