missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naratriptan hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Naratriptan hydrochloride is a selective 5-HT1 receptor subtype agonist and is a triptan drug that is used for the treatment of migraine headaches.
Specifications
Specifications
| Chemical Name or Material | Naratriptan hydrochloride |
| CAS | 143388-64-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H26ClN3O2S |
| Quantity | 10 mg |
| Synonym | GR-85548A hydrochloride |
| Solubility Information | DMSO : ≥ 35 mg/mL (94.10 mM) ∣H2O : 6.67 mg/mL (17.93 mM; Need ultrasonic) |
| SMILES | O=S(CCC1=CC2=C(NC=C2C3CCN(C)CC3)C=C1)(NC)=O.Cl |
| Molecular Weight (g/mol) | 371.93 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction