missing translation for 'onlineSavingsMsg'
Learn More

Naquotinib mesylate, MedChemExpress

Product Code. 16453696
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453696

Brand: Medchem Express HY19803

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Naquotinib mesylate (ASP8273 mesylate) is an orally available, mutant-selective and irreversible EGFR inhibitor; with IC50s of 8-33 nM toward EGFR mutants and 230 nM for EGFR.

Naquotinib mesylate (ASP8273 mesylate) is an orally available, mutant-selective and irreversible EGFR inhibitor

Specifications

Chemical Name or Material Naquotinib mesylate
CAS 1448237-05-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H46N8O6S
Quantity 25 mg
Synonym ASP8273 (mesylate)
Solubility Information DMSO : 12.5 mg/mL (18.97 mM; Need ultrasonic and warming)
SMILES O=C(C1=NC(CC)=C(O[C@H]2CN(C(C=C)=O)CC2)N=C1NC3=CC=C(N4CCC(N5CCN(C)CC5)CC4)C=C3)N.CS(=O)(O)=O
Molecular Weight (g/mol) 658.81
Formula Weight 658.81
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.