missing translation for 'onlineSavingsMsg'
Learn More

Naquotinib mesylate, MedChemExpress

Product Code. 16463857
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463857 50 mg 50mg
16471869 1 mg 1mg
16460378 5 mg 5mg
16453696 25 mg 25mg
16417918 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16463857 Supplier Medchem Express Supplier No. HY19803
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Naquotinib mesylate (ASP8273 mesylate) is an orally available, mutant-selective and irreversible EGFR inhibitor; with IC50s of 8-33 nM toward EGFR mutants and 230 nM for EGFR.

Naquotinib mesylate (ASP8273 mesylate) is an orally available, mutant-selective and irreversible EGFR inhibitor

Specifications

Chemical Name or Material Naquotinib mesylate
CAS 1448237-05-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H46N8O6S
Quantity 50 mg
Synonym ASP8273 (mesylate)
Solubility Information DMSO : 12.5 mg/mL (18.97 mM; Need ultrasonic and warming)
SMILES O=C(C1=NC(CC)=C(O[C@H]2CN(C(C=C)=O)CC2)N=C1NC3=CC=C(N4CCC(N5CCN(C)CC5)CC4)C=C3)N.CS(=O)(O)=O
Molecular Weight (g/mol) 658.81
Formula Weight 658.81
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.