missing translation for 'onlineSavingsMsg'
Learn More

Napsin A Antibody (NAPSA/3307) - Azide and BSA Free, Novus Biologicals™

Product Code. 18356242 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356242 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356242 Supplier Novus Biologicals Supplier No. NBP308377

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Napsin A Monoclonal specifically detects Napsin A in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Napsin A
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone NAPSA/3307
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02
Host Species Mouse
Immunogen Recombinant human Napsin A protein fragment (aa189-299) (exact sequence is proprietary)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 9476
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.