missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Napsin A Antibody (CL12954), Novus Biologicals™
Mouse Monoclonal Antibody
£338.00 - £482.00
Specifications
| Antigen | Napsin A |
|---|---|
| Clone | CL12954 |
| Dilution | Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628754
|
Novus Biologicals
NBP3-18551-100ul |
100 μg |
£482.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683435
|
Novus Biologicals
NBP3-18551-25ul |
25 μg |
£338.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Napsin A Monoclonal antibody specifically detects Napsin A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Napsin A | |
| Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02 | |
| This antibody was developed against a sythetic peptide for O96009. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL12954 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cellular Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 9476 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title