missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPSA, Mouse anti-Human, Clone: 2B1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009476-M07.100ug
This item is not returnable.
View return policy
Description
Sequence: LIRIPLHRVQPGRRTLNLLRGWREPAELPKLGAPSPGDKPIFVPLSNYRDVQYFGEIGLGTPPQNFTVAFDTGSSNLWVPSRRCHFFSVPCWLHHRFDPKASSSFQANGTKFAIQYGTGRVDGILSEDKLTIGGIKGASVIFGEALWEPSLVFAFAHFDGILGLGFPILSVEGVRPPMDVLVEQGLLDKPVFSFYLNRDPEEPDGGELVLGGSDPAHYIPPLTFVPVTVPAYWQIHMERVKVGPGLTLCAKGCAAILDTGTSLITGPTEEIRALHAAIGGIPLLAGEYIILCSEIPKLPAVSFLLGGVWFNLTAHDYVIQTTRNGVRLCLSGFQALDVPPPAGPFWILGDVFLGTYVAVFDRGDMKSSARVGLARARTRGADLGWGETAQAQFPGSpecifications
| NAPSA | |
| Monoclonal | |
| Unconjugated | |
| napsin A aspartic peptidase | |
| KAP/Kdap/NAP1/NAPA/SNAPA | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2B1 | |
| In 1x PBS, pH 7.4 | |
| BC017842 | |
| NAPSA | |
| NAPSA (AAH17842, 26 a.a. ∼ 420 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9476 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction