missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAPSA Monoclonal Antibody (10C4B8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548669
This item is not returnable.
View return policy
Description
NAPSA Monoclonal Antibody for Western Blot, IHC (P), ELISA
Napsin 1 is an aspartic protease that is generally expressed in the lung, but also present in kidney and in metastatic carcinomas throughout the body. Napsin 1 processes surfactant protein B in healthy pulmonary and renal tissues, but is abnormally regulated when carcinogenesis occurs. Thus, many researchers have found napsin evaluation to be useful in identifying several cancers, and differentiating primary tumors from metastases. When combined with thyroid transcription factor analysis, napsin can be used as a specific biomarker for differentiation of carcinoma types. Napsin has also been associated with pulmonary alveolar proteinosis as well as in protein catabolism in renal tubules.
Specifications
| NAPSA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O96009 | |
| NAPSA | |
| Purified recombinant fragment of human NAPSA (AA: 20-158) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 10C4B8 | |
| Unconjugated | |
| NAPSA | |
| asp 4; ASP4; Aspartyl protease 4; KAP; Kdap; KDAP-1; kidney-derived aspartic protease-like protein; Nap; NAP1; NAPA; Napsa; napsin A aspartic peptidase; napsin-1; Napsin-A; pronapsin; pronapsin A; SNAPA; TA01/TA02 | |
| Mouse | |
| Protein G | |
| RUO | |
| 60575, 9476 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction