missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPRT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35172-100ul
This item is not returnable.
View return policy
Description
NAPRT1 Polyclonal antibody specifically detects NAPRT1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| NAPRT1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 2.4.2.11, FHA-HIT-interacting protein, FHIP, NAPRTase, nicotinate phosphoribosyltransferase, nicotinate phosphoribosyltransferase domain containing 1, Nicotinate phosphoribosyltransferase domain-containing protein 1, nicotinic acid phosphoribosyltransferase, PP3856 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 309-538 of human NAPRT1 (NP_660202.3).,, Sequence:, ELGYRAVGVRLDSGDLLQQAQEIRKVFRAAAAQFQVPWLESVLIVVSNNIDEEALARLAQEGSEVNVIGIGTSVVTCPQQPSLGGVYKLVAVGGQPRMKLTEDPEKQTLPGSKAAFRLLGSDGSPLMDMLQLAEEPVPQAGQELRVWPPGAQEPCTVRPAQVEPLLRLCLQQGQLCEPLPSLAESRALAQLSLSRLSPEHRRLRSPAQYQVVLSERLQALVNSLCAGQSP | |
| 100 μL | |
| Lipid and Metabolism | |
| 93100 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction