missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPRT1 Antibody (CL0366) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52924JF525
This item is not returnable.
View return policy
Description
NAPRT1 Monoclonal antibody specifically detects NAPRT1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| NAPRT1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| EC 2.4.2.11, FHA-HIT-interacting protein, FHIP, NAPRTase, nicotinate phosphoribosyltransferase, nicotinate phosphoribosyltransferase domain containing 1, Nicotinate phosphoribosyltransferase domain-containing protein 1, nicotinic acid phosphoribosyltransferase, PP3856 | |
| This antibody was developed using a recombinant protein derived from Q6XQN6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 93100 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0366 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction