missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naproxen Sodium, Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Naproxen Sodium
Specifications
Specifications
| Chemical Name or Material | Naproxen Sodium |
| CAS | 26159-34-2 |
| Analyte or Component Names | Naproxen Sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H13 O3 . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Naproxen Sodium, (2S)-2-(6-Methoxy-2-naphthyl)propanoic acid sodium salt, Aflaxen, Agilxen, Aleve, Anaprox, Anaprox DS, Anax, Apranax, Apraxin, Apronax, Axer, Dafloxen, Duk-F, Flanax, Flanax Forte, Gynestrel, Lefaine, Licorax, Miranax, Naprelan, Naprogesic, Naprosyn sodium, Naproxen sodium, Narocin, Primeral, RS 3560, Sodium (+)-6-methoxy-2-naphthalenepropionate, Sodium d-2-(6-methoxy-2-naphthyl)propionate, Sodium naprosyn, Sodium naproxen, Sutolin, Synflex, Synflex Forte, Uniflam, Xenobid, (alphaS)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid sodium salt (1:1), (S)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid sodium salt, (alphaS)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid sodium salt, L-(-)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid sodium salt |
| InChI Formula | InChI=1S/C14H14O3.Na/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10;/h3-9H,1-2H3,(H,15,16);/q;+1/p-1/t9-;/m0./s1 |
| SMILES | [Na+].COc1ccc2cc(ccc2c1)[C@H](C)C(=O)[O-] |
| IUPAC Name | sodium;(2S)-2-(6-methoxynaphthalen-2-yl)propanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction