missing translation for 'onlineSavingsMsg'
Learn More

Naproxen sodium, MedChemExpress

Product Code. p-200101522
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16436298

Brand: Medchem Express HY15030A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Naproxen sodium is a COX-1 and COX-2 inhibitor with IC50s of 8.72 and 5.15 μM, respectively in cell assay.

Naproxen sodium is a COX-1 and COX-2 inhibitor with IC50s of 8.72 and 5.15 μM, respectively in cell assay.

Specifications

Chemical Name or Material Naproxen sodium
CAS 26159-34-2
Color White
Purity Grade Notes Research
Molecular Formula C14H13NaO3
Quantity 10 g
Solubility Information H2O : 75 mg/mL (297.34 mM; Need ultrasonic and warming) ∣DMSO : 5 mg/mL (19.82 mM; Need ultrasonic)
SMILES C[C@H](C(O[Na])=O)C1=CC2=CC=C(OC)C=C2C=C1
Molecular Weight (g/mol) 252.24
Formula Weight 252.24
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.