missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naproxen-O-desmethyl, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | (2S)-2-(6-Hydroxynaphthalen-2-yl)propanoic Acid (O-Demethylnaproxen) |
| CAS | 52079-10-4 |
| Analyte or Component Names | (2S)-2-(6-Hydroxynaphthalen-2-yl)propanoic Acid (O-Demethylnaproxen) |
| Shipping Condition | +20°C |
| Molecular Formula | C13 H12 O3 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 2-Naphthaleneacetic acid, 6-hydroxy-α-methyl-, (S)-, (αS)-6-Hydroxy-α-methyl-2-naphthaleneacetic acid, (+)-2-(6-Hydroxy-2-naphthyl)propionic acid, (2S)-2-(6-Hydroxynaphthalen-2-yl)propanoic acid, (S)-2-(6-Hydroxy-2-naphthyl)propionic acid, (S)-6-O-Desmethylnaproxen, 6-Demethylnaproxen, 6-Desmethylnaproxen, 6-Hydroxynaproxen, 6-O-Demethylnaproxen, O-Desmethylnaproxen, 2-Naphthaleneacetic acid, 6-hydroxy-α-methyl-, (αS)-, Naproxen Imp. A (EP), Naproxen-O-desmethyl, (Naproxen Imp. A (EP), Naproxen Impurity A |
| InChI Formula | InChI=1S/C13H12O3/c1-8(13(15)16)9-2-3-11-7-12(14)5-4-10(11)6-9/h2-8,14H,1H3,(H,15,16)/t8-/m0/s1 |
| SMILES | C[C@H](C(=O)O)c1ccc2cc(O)ccc2c1 |
| IUPAC Name | (2S)-2-(6-hydroxynaphthalen-2-yl)propanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?