missing translation for 'onlineSavingsMsg'
Learn More

Naproxen, MedChemExpress

Product Code. p-200097486
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426298 10 g 10g
16418887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16426298 Supplier Medchem Express Supplier No. HY15030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Naproxen is a COX-1 and COX-2 inhibitor with IC50s of 8.72 and 5.15 μM, respectively in cell assay.

Naproxen is a COX-1 and COX-2 inhibitor with IC50s of 8.72 and 5.15 μM, respectively in cell assay.

Specifications

Chemical Name or Material Naproxen
CAS 22204-53-1
Color White
Purity Grade Notes Research
Molecular Formula C14H14O3
Quantity 10 g
Synonym (S)-Naproxen
Solubility Information DMSO : ≥ 100 mg/mL (434.29 mM) ∣H2O : 75 mg/mL (325.72 mM; Need ultrasonic and warming)
SMILES C[C@H](C(O)=O)C1=CC2=CC=C(OC)C=C2C=C1
Molecular Weight (g/mol) 230.26
Formula Weight 230.26
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.