missing translation for 'onlineSavingsMsg'
Learn More

Naproxen, Lipomed

Product Code. 30463090
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463090 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463090 Supplier Lipomed Supplier No. LPMNPR171850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Naproxen
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naproxen
CAS 22204-53-1
Analyte or Component Names Naproxen
Shipping Condition Room Temperature
Molecular Formula C14 H14 O3
Synonym (+)-(S)-Naproxen, (+)-2-(6-Methoxy-2-naphthyl)propionic acid, (+)-Naproxen, (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, (S)-(+)-Naproxen, 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES COc1ccc2cc(ccc2c1)[C@H](C)C(=O)O
IUPAC Name (2 S)-2-(6-methoxynaphthalen-2-yl)propanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 230.26
Quantity 50 mg
Formula Weight 230.0943 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.