missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human LIGHT / TNFSF14 Protein, Fc tag

Product Code. 16427849
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg

Product Code. 16427849

Brand: ACROBiosystems LITH5265100ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Tumor necrosis factor ligand superfamily member 14(LIGHT) is a tumor necrosis factor (TNF) superfamily ligand that regulates T cell immune responses by signaling through the herpes virus entry mediator (HVEM) and the lymphotoxin beta receptor (LTbetaR). LIGHT has emerged as a potent initiator of T cell co-stimulation signals effecting CTL-mediated tumor rejection, allograft rejection and graft versus host disease. Constitutive expression of LIGHT leads to tissue destruction and autoimmune-like disease syndromes.

This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 44.7 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP 003798.2
Molecular Weight (g/mol) 44.7 kDa
Quantity 100 μg
Source Human LIGHT, Fc tag (LIT-H5265) is expressed from human 293 cells (HEK293). It contains AA Asp 74 - Val 240 (Accession # O43557-1).
Immunogen Asp 74 - Val 240
Storage Requirements -20°C
Gene Alias CD258 antigen,CD258,HVEM-L,LIGHT,LTg,TNFSF14,TR2,HVEML
Common Name LIGHT
Conjugate Unconjugated
Species Human
Protein Tag Fc Tag
Buffer Tris with Glycine, Arginine and NaCl, pH7.5
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Immune Checkpoint Proteins
Purity or Quality Grade >85% as determined by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.